ENA250698865 Enamine

1-(1,4-diazepan-1-yl)-2-(2-methoxyphenyl)ethan-1-one

  • MDL number MFCD11164352
  • Cas Number : 1094240-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58888
Availability details for
Quantity
  • Availability
  • Add to Cart