ENA250698866 Enamine

3-fluoro-4-(2-fluorophenoxy)benzaldehyde

  • MDL number MFCD11156482
  • Cas Number : 1021233-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-58889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58889
Availability details for
Quantity
  • Availability
  • Add to Cart