ENA250698874 Enamine

2-(piperidin-4-yl)-1H-indole

  • MDL number MFCD05207607
  • Cas Number : 200714-50-7
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58916
Availability details for
Quantity
  • Availability
  • Add to Cart