ENA250698877 Enamine

N'-hydroxy-4-[(morpholin-4-yl)methyl]benzene-1-carboximidamide

  • Cas Number : 926198-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58919
Availability details for
Quantity
  • Availability
  • Add to Cart