ENA250698878 Enamine

4-(2-fluorophenoxymethyl)benzonitrile

  • MDL number MFCD09933637
  • Cas Number : 1016780-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58920

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58920
Availability details for
Quantity
  • Availability
  • Add to Cart