ENA250698879 Enamine

4-[(2-fluorophenoxy)methyl]-N'-hydroxybenzene-1-carboximidamide

  • Cas Number : 1016853-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58921
Availability details for
Quantity
  • Availability
  • Add to Cart