ENA250698880 Enamine

2-amino-2,3-diphenylpropanoic acid

  • MDL number MFCD02662655
  • Cas Number : 56594-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 264...266
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58922

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58922
Availability details for
Quantity
  • Availability
  • Add to Cart