ENA250698882 Enamine

4-(piperidine-1-sulfonyl)phenol

  • MDL number MFCD02709555
  • Cas Number : 99986-71-7
  • Molecular Weight : 241.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58925
Availability details for
Quantity
  • Availability
  • Add to Cart