ENA250698888 Enamine

4,4-dimethyl-1,2,3,4-tetrahydronaphthalen-1-one

  • MDL number MFCD00089606
  • Cas Number : 2979-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 14-Dec
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36504
Availability details for
Quantity
  • Availability
  • Add to Cart