ENA250698890 Enamine

2-amino-N-(butan-2-yl)propanamide

  • MDL number MFCD09937270
  • Cas Number : 1101928-01-1
  • Molecular Weight : 144.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58933
Availability details for
Quantity
  • Availability
  • Add to Cart