ENA250698894 Enamine

1-(1,4-diazepan-1-yl)-2-(3-fluorophenyl)ethan-1-one

  • MDL number MFCD09813907
  • Cas Number : 1016713-62-4
  • Molecular Weight : 236.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58937
Availability details for
Quantity
  • Availability
  • Add to Cart