ENA250698906 Enamine

5-(2-fluorophenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD09736685
  • Cas Number : 7248-25-1
  • Molecular Weight : 208.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58969
Availability details for
Quantity
  • Availability
  • Add to Cart