ENA250698909 Enamine

6-amino-1-[(4-chlorophenyl)methyl]-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD12577879
  • Cas Number : 1153515-25-3
  • Molecular Weight : 286.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58973
Availability details for
Quantity
  • Availability
  • Add to Cart