ENA250698910 Enamine

[(3-chloro-4-cyanophenyl)carbamoyl]formic acid

  • MDL number MFCD12171706
  • Cas Number : 1153559-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58974
Availability details for
Quantity
  • Availability
  • Add to Cart