ENA250698911 Enamine

5-(2,4-dimethoxyphenyl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD05182324
  • Cas Number : 502546-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58975
Availability details for
Quantity
  • Availability
  • Add to Cart