ENA250698914 Enamine

2-(4-chlorophenyl)-2-(morpholin-4-yl)ethan-1-amine

  • MDL number MFCD08437446
  • Cas Number : 866782-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58978

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58978
Availability details for
Quantity
  • Availability
  • Add to Cart