ENA250698915 Enamine

3-{[2-(propan-2-yloxy)ethoxy]methyl}aniline

  • MDL number MFCD09806390
  • Cas Number : 1016711-52-6
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58979
Availability details for
Quantity
  • Availability
  • Add to Cart