ENA250698920 Enamine

N-[4-(4-methoxyphenyl)butan-2-ylidene]hydroxylamine

  • Cas Number : 52271-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-58990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58990
Availability details for
Quantity
  • Availability
  • Add to Cart