ENA250698924 Enamine

5-amino-1-[(3,4-dichlorophenyl)methyl]-1,2-dihydropyridin-2-one

  • MDL number MFCD00139933
  • Cas Number : 339009-11-9
  • Molecular Weight : 269.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58994
Availability details for
Quantity
  • Availability
  • Add to Cart