ENA250698925 Enamine

(adamantan-1-yl)methyl N-ethylcarbamate

  • Cas Number : 194801-48-4
  • Molecular Weight : 237.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44262
Availability details for
Quantity
  • Availability
  • Add to Cart