ENA250698926 Enamine

1-(2-chlorophenyl)-1H-1,2,3-triazole-5-carboxylic acid

  • MDL number MFCD11559207
  • Cas Number : 101870-66-0
  • Molecular Weight : 223.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58999
Availability details for
Quantity
  • Availability
  • Add to Cart