ENA250698927 Enamine

3-chloro-2-hydrazinylpyridine

  • MDL number MFCD01936479
  • Cas Number : 22841-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44833

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-44833
pictograms GHS07,GHS08
Availability details for
Quantity
  • Availability
  • Add to Cart