ENA250698930 Enamine

oxane-4-sulfonyl chloride

  • MDL number MFCD07787616
  • Cas Number : 338453-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-35
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59004

Safety Information

signalword Danger
MSDS https://www.enaminestore.com/catalog/EN300-59004
pictograms GHS05
hstatements EUH029,H314
Availability details for
Quantity
  • Availability
  • Add to Cart