ENA250698933 Enamine

2-amino-N-[(furan-2-yl)methyl]propanamide

  • MDL number MFCD09950370
  • Cas Number : 1164109-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59008
Availability details for
Quantity
  • Availability
  • Add to Cart