ENA250698934 Enamine

3-methyl-2-[(oxolan-2-yl)methoxy]butanoic acid

  • MDL number MFCD09941758
  • Cas Number : 1016785-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59009
Availability details for
Quantity
  • Availability
  • Add to Cart