ENA250698936 Enamine

[2-(dimethylamino)ethyl][(3-fluorophenyl)methyl]amine

  • MDL number MFCD09049235
  • Cas Number : 926240-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59014

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59014
Availability details for
Quantity
  • Availability
  • Add to Cart