ENA250698947 Enamine

2-(3,4-difluorophenoxy)propanoic acid

  • MDL number MFCD08444523
  • Cas Number : 923113-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59037
Availability details for
Quantity
  • Availability
  • Add to Cart