ENA250698949 Enamine

5-(pyridin-4-yl)-4-(2,2,2-trifluoroethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD11207291
  • Cas Number : 1094377-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59039
Availability details for
Quantity
  • Availability
  • Add to Cart