ENA250698953 Enamine

3-phenyl-1,2,4-thiadiazole-5-thiol

  • MDL number MFCD00086459
  • Cas Number : 20069-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49426

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49426
Availability details for
Quantity
  • Availability
  • Add to Cart