ENA250698957 Enamine

2-(4-chlorophenoxy)-2-phenylacetic acid

  • MDL number MFCD09707683
  • Cas Number : 119061-18-6
  • Molecular Weight : 262.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59047
Availability details for
Quantity
  • Availability
  • Add to Cart