ENA250698958 Enamine

methyl 3-tert-butyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD00652984
  • Cas Number : 152307-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59050
Availability details for
Quantity
  • Availability
  • Add to Cart