ENA250698961 Enamine

6-(ethylsulfanyl)pyridin-3-amine

  • MDL number MFCD10007807
  • Cas Number : 52025-15-7
  • Molecular Weight : 154.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59053
Availability details for
Quantity
  • Availability
  • Add to Cart