ENA250698967 Enamine

1-fluoro-2-isothiocyanato-4-methylbenzene

  • MDL number MFCD05864298
  • Cas Number : 832113-98-1
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59060
Availability details for
Quantity
  • Availability
  • Add to Cart