ENA250698975 Enamine

4-(2,5-dichlorophenoxy)-3-fluoroaniline

  • MDL number MFCD11193294
  • Cas Number : 1039869-08-3
  • Molecular Weight : 272.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59074
Availability details for
Quantity
  • Availability
  • Add to Cart