ENA250698978 Enamine

N-(4-chlorophenyl)-1-methylpiperidin-4-amine

  • MDL number MFCD01653850
  • Cas Number : 36796-53-9
  • Molecular Weight : 224.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59077
Availability details for
Quantity
  • Availability
  • Add to Cart