ENA250698980 Enamine

4-(4-fluorophenyl)-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD09746481
  • Cas Number : 834885-06-2
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59080
Availability details for
Quantity
  • Availability
  • Add to Cart