ENA250698988 Enamine

[(2,4-difluorophenyl)methyl][2-(dimethylamino)ethyl]amine

  • MDL number MFCD09046101
  • Cas Number : 926258-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59091

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59091
Availability details for
Quantity
  • Availability
  • Add to Cart