ENA250698989 Enamine

[(3,4-difluorophenyl)methyl][2-(dimethylamino)ethyl]amine

  • MDL number MFCD09047827
  • Cas Number : 926270-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59092

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59092
Availability details for
Quantity
  • Availability
  • Add to Cart