ENA250698992 Enamine

4-methyl-2-[(4-methylbenzene)sulfonamido]pentanoic acid

  • MDL number MFCD00067467
  • Cas Number : 67368-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59096
Availability details for
Quantity
  • Availability
  • Add to Cart