ENA250699002 Enamine

4-(methylsulfanyl)-2-[(quinolin-2-yl)formamido]butanoic acid

  • MDL number MFCD13356161
  • Cas Number : 170488-18-3
  • Molecular Weight : 304.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59129
Availability details for
Quantity
  • Availability
  • Add to Cart