ENA250699011 Enamine

3-(2,4-dichlorophenoxy)propan-1-amine

  • MDL number MFCD09883063
  • Cas Number : 50911-63-2
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59144
Availability details for
Quantity
  • Availability
  • Add to Cart