ENA250699013 Enamine

N-methylethane-1-sulfonamide

  • MDL number MFCD06042345
  • Cas Number : 6601-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 7-Mar
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59146
Availability details for
Quantity
  • Availability
  • Add to Cart