ENA250699040 Enamine

4-(2-fluorophenyl)-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD09746485
  • Cas Number : 1095051-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59193
Availability details for
Quantity
  • Availability
  • Add to Cart