ENA250699046 Enamine

1,3-diphenylpropan-2-ol

  • MDL number MFCD00022047
  • Cas Number : 5381-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59200
Availability details for
Quantity
  • Availability
  • Add to Cart