ENA250699048 Enamine

3-{[(3-methylcyclohexyl)oxy]methyl}aniline

  • MDL number MFCD11196406
  • Cas Number : 1037157-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59203
Availability details for
Quantity
  • Availability
  • Add to Cart