ENA250699052 Enamine

1-[(2-bromoethyl)sulfanyl]-4-chlorobenzene

  • MDL number MFCD09948099
  • Cas Number : 13290-31-8
  • Molecular Weight : 251.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59208
Availability details for
Quantity
  • Availability
  • Add to Cart