ENA250699054 Enamine

N-[2-(3-fluorophenyl)ethyl]-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD12139944
  • Cas Number : 1153979-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59212
Availability details for
Quantity
  • Availability
  • Add to Cart