ENA250699057 Enamine

3-amino-1-(2-fluoro-5-methylphenyl)thiourea

  • MDL number MFCD11192321
  • Cas Number : 1037153-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-59216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59216
Availability details for
Quantity
  • Availability
  • Add to Cart