ENA250699058 Enamine

(2-fluoro-5-methylphenyl)thiourea

  • MDL number MFCD11202366
  • Cas Number : 1038356-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59217
Availability details for
Quantity
  • Availability
  • Add to Cart