ENA250699059 Enamine

methyl 4-carbamothioylbenzoate

  • MDL number MFCD09759040
  • Cas Number : 80393-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59218

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59218
Availability details for
Quantity
  • Availability
  • Add to Cart